Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAM-1/CD226 Antibody (2770B), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB305377 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305377 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305377 Supplier Novus Biologicals Supplier No. FAB10988K
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

DNAM-1/CD226 Monoclonal antibody specifically detects DNAM-1/CD226 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen DNAM-1/CD226
Applications Flow Cytometry
Classification Monoclonal
Clone 2770B
Conjugate DyLight 488
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1
Host Species Rabbit
Immunogen Human embryonic kidney cell HEK293-derived human DNAM-1/CD226 protein, Glu19-Asn247, Accession # Q15762
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10666
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.