Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAM-1/CD226 Rabbit anti-Human, Clone: 102, Novus Biologicals™
SDP

Catalog No. NB020626 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020626 50 μg
NB020625 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB020626 Supplier Novus Biologicals Supplier No. NBP28957150UL

Rabbit Monoclonal Antibody

DNAM-1/CD226 Monoclonal specifically detects DNAM-1/CD226 in Human samples. It is validated for ELISA.

Specifications

Antigen DNAM-1/CD226
Applications ELISA
Classification Monoclonal
Clone 102
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1
Host Species Rabbit
Immunogen Recombinant Human DNAM-1/CD226 protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10666
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.