Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAM-1/CD226 Rabbit anti-Human, Clone: 102, Novus Biologicals™

Rabbit Monoclonal Antibody
$366.00 - $524.00
Specifications
| Antigen | DNAM-1/CD226 |
|---|---|
| Clone | 102 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
Description
DNAM-1/CD226 Monoclonal specifically detects DNAM-1/CD226 in Human samples. It is validated for ELISA.Specifications
| DNAM-1/CD226 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1 | |
| Recombinant Human DNAM-1/CD226 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 102 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Innate Immunity, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 10666 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title