Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DNAPac™ RP Columns

Catalog No. 03100097
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
3 mm
21.2 mm
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
03-100-097 150 mm 21.2 mm
DX088919 100 mm 3 mm
DX088920 50 mm 3 mm
DX088921 10 mm 3 mm
DX088923 100 mm 2.1 mm
DX088924 50 mm 2.1 mm
DX088925 10 mm 2.1 mm
03-254-124 250 mm 2.1 mm
03-100-098 10 mm 21.2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 03-100-097 Supplier Thermo Scientific™ Supplier No. 0809221521232
Add to Cart
Add to Cart
Request a Quote

Get high resolution LC and LC-MS separation of oligonucleotides, including double-stranded (ds) DNA/RNA fragments, using these DNAPac reversed phase columns.

Achieve superior reversed phase oligonucleotide separations using the Thermo Scientific™ DNAPac™ RP HPLC columns. The unique chemistry is designed for analysis of oligonucleotides and double-stranded (ds) DNA/RNA fragments using LC-UV or LC-MS. The column chemistry provides excellent performance under a broad range of pH, temperature, and mobile phase compositions. In addition, the wide pore size of the resin provides excellent separation of large double-stranded nucleic acids up to 10k base pairs.

DNAPac RP reversed phase (RP) column is based on spherical wide-pore 4 μm polymer resin specifically designed for separation of oligonucleotides and DNA/RNA fragments. The stationary phase is highly stable to extreme pH (0-14), and temperature (5-110°C) and fully compatible with mass spectrometry friendly organic solvents and ion-pair reagents such as triethylamine (TEA) and hexylamine (HA). The wide pore size of polymer resin enables efficient separation of small to large nucleic acids with low carry-over. The column may be operated in both HPLC and UHPLC instruments, producing high resolution at high throughput for oligonucleotides and resolves both small and very large dsDNA segments:

  • Designed for ion-pair reversed-phase (IP-RP) LC and LC/MS separations of both oligonucleotides and double-stranded nucleic acids
  • High resolution and high throughput
  • Excellent MS compatibility
  • Wide operating pH range (0-14) and high temperature stability (up to 110°C)
  • Biocompatible hardware to eliminate analyte absorption
TRUSTED_SUSTAINABILITY

Specifications

Column Format Preparative column, standard
Diameter (Metric) 21.2 mm
Flow Rate 4 to 8 mL/min
Length (Metric) 150 mm
Max. Pressure 2900 psi (200 bar)
Particle Size 4 μm
pH 0 to 14
Quantity Each
Temperature up to 110°C
Stationary Phase Proprietary
Product Line DNAPac
Packing Material 4 µm polymer Resin
Column Type Reversed Phase
Phase Reversed Phase
Material DVB (Divinylbenzene) Polymer
Technique Reversed Phase HPLC
Product Type Preparative Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.