Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNase II Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB044420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044420 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044420 Supplier Novus Biologicals Supplier No. NBP297515DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNase II Polyclonal specifically detects DNase II in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DNase II
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Acid DNase, Deoxyribonuclease II alpha, deoxyribonuclease II, lysosomal, deoxyribonuclease-2-alpha, DNase II alpha, DNase II, lysosomal, DNASE2A, DNL, DNL2, EC 3.1.22.1, Lysosomal DNase II, R31240_2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DNase II fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1777
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.