Learn More
Description
Specifications
Specifications
| Antigen | DNASE1L3 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 |
| Formulation | PBS pH7, 20% Glycerol with 0.025% Proclin 300 |
| Gene Accession No. | Q13609 |
| Gene Alias | deoxyribonuclease gamma, deoxyribonuclease I-like 3Liver and spleen DNase, deoxyribonuclease I-like III, DNAS1L3LSDDHP2, DNase gamma, DNase I homolog protein 2, DNase I homolog protein DHP2, DNase I-like 3, EC 3.1.21.-, EC 3.1.21.1, LS-DNase |
| Gene Symbols | DNASE1L3 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
