Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNCIC1 Antibody (74.1), Novus Biologicals™
SDP

Catalog No. NB300726 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300726 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300726 Supplier Novus Biologicals Supplier No. NB300726
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNCIC1 Monoclonal specifically detects DNCIC1 in Human, Mouse, Rat, Drosophila, Mammal, Xenopus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen DNCIC1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 74.1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Flow Cytometry 1 - 2 ug, Immunocytochemistry/Immunofluorescence 2 - 4 ug/ml, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500
Gene Accession No. O14576
Gene Alias cytoplasmic dynein 1 intermediate chain 1, Cytoplasmic dynein intermediate chain 1, DNCI1cytoplasmic, intermediate polypeptide 1, DNCIC1DH IC-1, Dynein intermediate chain 1, cytosolic, dynein, cytoplasmic 1, intermediate chain 1
Gene Symbols DYNC1I1
Host Species Mouse
Immunogen Purified bovine brain cytoplasmic dynein.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1780
Test Specificity Detects two isoforms of the 74kDa polypeptide of cytoplasmic Dynein.
Target Species Human, Mouse, Rat, Drosophila, Mammalia, Xenopus species
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.