Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMBP Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB291121 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291121 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291121 Supplier Novus Biologicals Supplier No. NBP171830APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNMBP Polyclonal antibody specifically detects DNMBP in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen DNMBP
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias dynamin binding protein, dynamin-binding protein, KIAA1010ARHGEF36, Scaffold protein Tuba, TUBA
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 75 and 125 of human Scaffold Protein TUBA using the numbering given in entry NP_056036.1 (GeneID 23268).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23268
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.