Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNMT1, Ac-4K Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $498.79
Specifications
Antigen | DNMT1, Ac-4K |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Dot Blot, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABE1956-25UG
|
MilliporeSigma
ABE195625UG |
25 μg |
Each for $192.35
|
|
|||||
ABE1956-100
|
MilliporeSigma
ABE1956100UG |
100 μg |
Each for $498.79
|
|
|||||
Description
Anti-DNMT1, Ac-4K, Cat. No. ABE1956, is a highly specific rabbit polyclonal antibody that targets DNA (cytosine-5)-methyltransferase 1 (DNMT1) and has been tested for use in Dot Blot, Immunocytochemistry, and Western Blotting.Specifications
DNMT1, Ac-4K | |
Dot Blot, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Epigenetics & Nuclear Function | |
Purified rabbit polyclonal antibody in PBS with 0.02% sodium azide and 50% glycerol. | |
NP_001370 | |
KLH-conjugated linear peptide corresponding to 11 amino acids from the C-terminal half with acetylated K1111, 1113, 1115, and 1117. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
Antibodies |
Please refer to lot specific datasheet. | |
Polyclonal | |
Purified | |
RUO | |
Human | |
DNA (cytosine-5)-methyltransferase 1;EC: 2.1.1.37;CXXC-type zinc finger protein 9;DNA methyltransferase HsaI;DNA MTase HsaI;M.HsaI;MCMT | |
DNMT1;AIM;CXXC9;DNMT | |
IgG | |
Affinity Purified | |
This rabbit polyclonal antibody specifically detects DNA (cytosine-5)-methyltransferase 1 acetylated on four lysine residues from the C-terminal half containing six tandem repeats of K-G. | |
∽200 kDa observed; 183.16 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title