Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT1 Antibody (1A9), Novus Biologicals™
SDP

Catalog No. NB297749 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297749 100 μL
NB297748 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297749 Supplier Novus Biologicals Supplier No. NBP326614100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

DNMT1 Monoclonal antibody specifically detects DNMT1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen DNMT1
Applications ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 1A9
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992
Host Species Rabbit
Immunogen A synthesized peptide derived from Human DNMT1 [UniProt P26358]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Chromatin Research, Core ESC Like Genes, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1786
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.