Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNMT1 Antibody (1A9), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | DNMT1 |
---|---|
Clone | 1A9 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
DNMT1 Monoclonal antibody specifically detects DNMT1 in Human samples. It is validated for ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
DNMT1 | |
ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992 | |
A synthesized peptide derived from Human DNMT1 [UniProt P26358] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
1A9 | |
ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Chromatin Research, Core ESC Like Genes, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1786 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title