Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT1 Antibody (4G11-C7), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP259344UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259344UV 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259344UV Supplier Novus Biologicals Supplier No. NBP259344UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNMT1 Monoclonal specifically detects DNMT1 in Human, Mouse, Fish, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen DNMT1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 4G11-C7
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992
Gene Symbols DNMT1
Host Species Mouse
Immunogen Raised against a partial protein corresponding to amino acids 620-950 of human DNMT1
Purification Method Protein G purified
Quantity 100 μL
Research Discipline Angiogenesis, Cancer, Chromatin Modifiers, Chromatin Research, Core ESC Like Genes, DNA Methyltransferases, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1786
Test Specificity Detects 180kDa. It will cross-react with mouse DNMT1.
Target Species Human, Mouse, Fish, Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.