Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT1 Antibody (60B1220.1) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013770 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013770 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013770 Supplier Novus Biologicals Supplier No. NBP280697

Mouse Monoclonal Antibody

DNMT1 Monoclonal specifically detects DNMT1 in Human, Mouse, Rat, Porcine, Bovine, Sheep samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Immunohistochemistry Free-Floating, CyTOF-ready, Knockdown Validated.

Specifications

Antigen DNMT1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown
Classification Monoclonal
Clone 60B1220.1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 0.1 - 0.5 ug/ml, Simple Western 1:100, Flow Cytometry : 1 - 5 ug/ml, Immunohistochemistry : 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:10. Use reported in scientific literature (PMID 24386225), Immunoprecipitation : 1:10 - 1:500, Immunohistochemistry-Paraffin : 1:50 - 1:500, Immunohistochemistry-Frozen : 1:500, Flow (Intracellular) reported in scientific literature (PMID 24578214), Immunohistochemistry Free-Floating : 1:500, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000, CyTOF-ready, Knockdown Validated
Formulation PBS
Gene Alias AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992
Gene Symbols DNMT1
Host Species Mouse
Immunogen This antibody was raised against a synthetic peptide corresponding to amino acids 637-650 (EKDDREDKENAFKR) of human Dnmt1 (Genbank Accession No. NP_001370).
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Chromatin Research, Core ESC Like Genes, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1786
Test Specificity Rat reactivity reported in scientific literature (PMID: 19924110). Porcine reactivity reported in scientific literature (PMID: 23808878). Sheep reactivity reported in scientific literature (PMID: 20413592). Bovine reactivity reported in scientific literature (PMID:27384909).
Target Species Human, Mouse, Rat, Pig, Bovine, Sheep
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.