Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNMT1 Antibody (60B1220.1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 119 publications
Supplier: Novus Biologicals NB10056519
Description
DNMT1 Monoclonal specifically detects DNMT1 in Human, Mouse, Rat, Porcine, Bovine, Sheep samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Immunohistochemistry Free-Floating, CyTOF-ready, Knockdown Validated.Specifications
| DNMT1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 0.1-0.5 ug/ml, Simple Western 1:100, Flow Cytometry 1 - 5 ug/mL, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:10. Use reported in scientific literature (PMID 24386225), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen 1:500, Flow (Intracellular) reported in scientific literature (PMID 24578214), Immunohistochemistry Free-Floating 1:500, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000, CyTOF-ready, KnockDown Validated reported in scientific literature (PMID 31799621) | |
| AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown | |
| 60B1220.1 | |
| Unconjugated | |
| P26358 | |
| DNMT1 | |
| This antibody was raised against a synthetic peptide corresponding to amino acids 637-650 (EKDDREDKENAFKR) of human Dnmt1 (Genbank Accession No. NP_001370). | |
| 0.1 mg | |
| Chromatin Modifiers, Chromatin Research, Core ESC Like Genes, DNA Methyltransferases, Epigenetics, Stem Cell Markers, Transcription Factors and Regulators | |
| 1786 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction