Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNMT1 Antibody (60B1220.1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 119 publications
$244.50 - $584.50
Specifications
| Antigen | DNMT1 |
|---|---|
| Clone | 60B1220.1 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.1-0.5 ug/ml, Simple Western 1:100, Flow Cytometry 1 - 5 ug/mL, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:10. Use reported in scientific literature (PMID 24386225), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen 1:500, Flow (Intracellular) reported in scientific literature (PMID 24578214), Immunohistochemistry Free-Floating 1:500, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000, CyTOF-ready, KnockDown Validated reported in scientific literature (PMID 31799621) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056519T
![]() |
Novus Biologicals
NB10056519SS |
0.025 mg |
Each for $244.50
|
|
|||||
NB10056519
![]() |
Novus Biologicals
NB10056519 |
0.1 mg |
Each for $584.50
|
|
|||||
Description
DNMT1 Monoclonal specifically detects DNMT1 in Human, Mouse, Rat, Porcine, Bovine, Sheep samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Immunohistochemistry Free-Floating, CyTOF-ready, Knockdown Validated.Specifications
| DNMT1 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown | |
| Unconjugated | |
| Mouse | |
| Chromatin Modifiers, Chromatin Research, Core ESC Like Genes, DNA Methyltransferases, Epigenetics, Stem Cell Markers, Transcription Factors and Regulators | |
| P26358 | |
| 1786 | |
| This antibody was raised against a synthetic peptide corresponding to amino acids 637-650 (EKDDREDKENAFKR) of human Dnmt1 (Genbank Accession No. NP_001370). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 60B1220.1 | |
| Western Blot 0.1-0.5 ug/ml, Simple Western 1:100, Flow Cytometry 1 - 5 ug/mL, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:10. Use reported in scientific literature (PMID 24386225), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen 1:500, Flow (Intracellular) reported in scientific literature (PMID 24578214), Immunohistochemistry Free-Floating 1:500, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000, CyTOF-ready, KnockDown Validated reported in scientific literature (PMID 31799621) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig | |
| AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992 | |
| DNMT1 | |
| IgG1 κ | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title