Learn More
DNMT1 Antibody (60B1220.1) - BSA Free, Novus Biologicals™
SDP
Mouse Monoclonal Antibody has been used in 119 publications
Supplier:  Novus Biologicals NB10056519SS
 View more versions of this product
Catalog No. NB10056519T
Only null left
Add to Cart
Description
Description
DNMT1 Monoclonal specifically detects DNMT1 in Human, Mouse, Rat, Porcine, Bovine, Sheep samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Immunohistochemistry Free-Floating, CyTOF-ready, Knockdown Validated.
Specifications
Specifications
DNMT1
Monoclonal
1.0 mg/mL
Western Blot 0.1-0.5 ug/ml, Simple Western 1:100, Flow Cytometry 1 - 5 ug/mL, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:10. Use reported in scientific literature (PMID 24386225), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen 1:500, Flow (Intracellular) reported in scientific literature (PMID 24578214), Immunohistochemistry Free-Floating 1:500, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000, CyTOF-ready, KnockDown Validated reported in scientific literature (PMID 31799621)
AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992
Mouse
Protein G purified
RUO
Primary
Human, Mouse, Rat, Pig
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown
60B1220.1
Unconjugated
P26358
DNMT1
This antibody was raised against a synthetic peptide corresponding to amino acids 637-650 (EKDDREDKENAFKR) of human Dnmt1 (Genbank Accession No. NP_001370).
0.025 mg
Chromatin Modifiers, Chromatin Research, Core ESC Like Genes, DNA Methyltransferases, Epigenetics, Stem Cell Markers, Transcription Factors and Regulators
1786
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only