Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB363084 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363084 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363084 Supplier Novus Biologicals Supplier No. NB100392AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNMT1 Polyclonal specifically detects DNMT1 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen DNMT1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 700 and 750 of human DNA-Methyltranserase 1 using the numbering given in entry NP_001370.1 (GeneID 1786).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Chromatin Research, Core ESC Like Genes, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1786
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.