Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT1 Antibody (DNMT1/2061), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB386640 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB386640 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB386640 Supplier Novus Biologicals Supplier No. NBP308482AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNMT1 Monoclonal antibody specifically detects DNMT1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen DNMT1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DNMT1/2061
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992
Host Species Mouse
Immunogen Recombinant human DNMT1 protein fragment (around aa 767-912) (exact sequence is proprietary) (Uniprot: P26358)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Chromatin Research, Core ESC Like Genes, Epigenetics, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1786
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.