Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dnmt2 Antibody (102B1259.2), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB200587C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200587C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB200587C Supplier Novus Biologicals Supplier No. NB200587C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dnmt2 Monoclonal specifically detects Dnmt2 in Human, Mouse, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Dnmt2
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 102B1259.2
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Gene Accession No. Q6ICS7
Gene Alias DMNT2, DNA (cytosine-5-)-methyltransferase 2, DNA (cytosine-5)-methyltransferase-like protein 2, DNA methyltransferase homolog HsaIIP, DNA methyltransferase-2, DNA MTase homolog HsaIIP, Dnmt2, EC 2.1.1, EC 2.1.1.29, M.HsaIIP, MHSAIIP, PuMet, RNMT1, tRNA (cytosine-5-)-methyltransferase, tRNA aspartic acid methyltransferase 1
Gene Symbols TRDMT1
Host Species Mouse
Immunogen This antibody was raised against synthetic peptides corresponding to mouse Dnmt2.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 1787
Test Specificity In mouse Dnmt2 transfected cell lysate, a 42 kDa band is oberseved.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.