Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dnmt2 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB057165 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB057165 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB057165 Supplier Novus Biologicals Supplier No. NBP298992FR

Rabbit Polyclonal Antibody

Dnmt2 Polyclonal specifically detects Dnmt2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Dnmt2
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DMNT2, DNA (cytosine-5-)-methyltransferase 2, DNA (cytosine-5)-methyltransferase-like protein 2, DNA methyltransferase homolog HsaIIP, DNA methyltransferase-2, DNA MTase homolog HsaIIP, Dnmt2, EC 2.1.1, EC 2.1.1.29, M.HsaIIP, MHSAIIP, PuMet, RNMT1, tRNA (cytosine-5-)-methyltransferase, tRNA aspartic acid methyltransferase 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human Dnmt2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 1787
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.