Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dnmt2 Antibody, Novus Biologicals™
SDP

Catalog No. NB200616SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200616SS 0.025 mg
NB200616 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB200616SS Supplier Novus Biologicals Supplier No. NB200616SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Dnmt2 Polyclonal specifically detects Dnmt2 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Dnmt2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Alias DMNT2, DNA (cytosine-5-)-methyltransferase 2, DNA (cytosine-5)-methyltransferase-like protein 2, DNA methyltransferase homolog HsaIIP, DNA methyltransferase-2, DNA MTase homolog HsaIIP, Dnmt2, EC 2.1.1, EC 2.1.1.29, M.HsaIIP, MHSAIIP, PuMet, RNMT1, tRNA (cytosine-5-)-methyltransferase, tRNA aspartic acid methyltransferase 1
Gene Symbols TRDMT1
Host Species Rabbit
Immunogen This antibody was raised against synthetic peptides corresponding amino acids 39-53 and 361-376 of mouse Dnmt2.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 1787
Test Specificity Cross-reacts with Human and Mouse. Not yet tested in other species.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.