Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT3A Antibody (64B1446) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB12013888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12013888 0.1 mg
NB12013888T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB12013888 Supplier Novus Biologicals Supplier No. NB12013888
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNMT3A Monoclonal specifically detects DNMT3A in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry Whole-Mount, CyTOF.

Specifications

Antigen DNMT3A
Applications Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 64B1446
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Chromatin Immunoprecipitation reported in scientific literature (PMID 24623306), Flow Cytometry 1 ug per million cells, Immunohistochemistry reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml. Use reported in scientific literature (PMID 22134929), Immunohistochemistry-Frozen reported in scientific literature (PMID 22134929), Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 26507142), Chromatin Immunoprecipitation (ChIP), CyTOF-ready
Gene Accession No. Q9Y6K1
Gene Alias 64B1446 clone, DNA (cytosine-5-)-methyltransferase 3 alpha, DNA (cytosine-5)-methyltransferase 3A, DNA cytosine methyltransferase 3A2, DNA methyltransferase HsaIIIA, DNA MTase HsaIIIA, Dnmt3a, DNMT3A2, EC 2.1.1.37, M.HsaIIIA
Gene Symbols DNMT3A
Host Species Mouse
Immunogen This DNMT3A antibody was raised against bacteria expressed recombinant mouse Dnmt3a. The epitope was found to lie near the C-terminus (a.a. 705-908), see Chen et (2002) for details.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 1788
Test Specificity Cross-reacts with Human (weakly) and Mouse. Not yet tested in other species.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.