Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT3A Antibody (64B814.1), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N233216J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N233216J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N233216J646 Supplier Novus Biologicals Supplier No. NBP233216JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNMT3A Monoclonal specifically detects DNMT3A in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DNMT3A
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 64B814.1
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DNA (cytosine-5-)-methyltransferase 3 alpha, DNA (cytosine-5)-methyltransferase 3A, DNA cytosine methyltransferase 3A2, DNA methyltransferase HsaIIIA, DNA MTase HsaIIIA, Dnmt3a, DNMT3A2, EC 2.1.1.37, M.HsaIIIA
Gene Symbols DNMT3A
Host Species Mouse
Immunogen This DNMT3A Antibody (64B814.1) was raised against bacteria expressed HIS-tag recombinant mouse Dnmt3a.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 1788.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.