Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT3A Antibody (PCRP-DNMT3A-1E2) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB119209 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119209 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119209 Supplier Novus Biologicals Supplier No. NBP308483
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNMT3A Monoclonal specifically detects DNMT3A in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen DNMT3A
Applications Western Blot, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone PCRP-DNMT3A-1E2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1-2 ug/mL, Immunohistochemistry-Paraffin : 1-2 ug/mL, Protein Array
Formulation 10mM PBS
Gene Alias DNA (cytosine-5-)-methyltransferase 3 alpha, DNA (cytosine-5)-methyltransferase 3A, DNA cytosine methyltransferase 3A2, DNA Methyltransferase 3 Alpha, DNA methyltransferase HsaIIIA, DNA MTase HsaIIIA, Dnmt3a, DNMT3A2, EC 2.1.1.37, M.HsaIIIA, TBRS
Host Species Mouse
Immunogen Recombinant human full-length DNMT3A protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Chromatin Research, Epigenetics, Innate Immunity, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 1788
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.