Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT3B Antibody, Novus Biologicals™
SDP

Catalog No. NBP216206 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216206 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216206 Supplier Novus Biologicals Supplier No. NBP216206

Rabbit Polyclonal Antibody

DNMT3B Polyclonal antibody specifically detects DNMT3B in Mouse samples. It is validated for Western Blot.

Specifications

Antigen DNMT3B
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. O88509
Gene Alias DNA (cytosine-5-)-methyltransferase 3 beta, DNA (cytosine-5)-methyltransferase 3B, DNA methyltransferase HsaIIIB, DNA MTase HsaIIIB, Dnmt3b, EC 2.1.1.37, ICF, M.HsaIIIB
Gene Symbols DNMT3B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of mouse DNMT3B. The exact sequence is proprietary.
Molecular Weight of Antigen 97 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, DNA Repair, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 1789
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.