Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT3B Antibody, Novus Biologicals™
SDP

Catalog No. NBP259291 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259291 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259291 Supplier Novus Biologicals Supplier No. NBP259291
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNMT3B Polyclonal specifically detects DNMT3B in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DNMT3B
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, ChIP Assay
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:1000, Chromatin Immunoprecipitation (ChIP)
Formulation PBS with 0.05% Sodium Azide and 0.05% ProClin 300
Gene Alias DNA (cytosine-5-)-methyltransferase 3 beta, DNA (cytosine-5)-methyltransferase 3B, DNA methyltransferase HsaIIIB, DNA MTase HsaIIIB, Dnmt3b, EC 2.1.1.37, ICF, M.HsaIIIB
Gene Symbols DNMT3B
Host Species Rabbit
Immunogen The exact immunogen is proprietary information.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer, Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, DNA Repair, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 1789
Target Species Human
Content And Storage Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.