Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT3B Antibody, Novus Biologicals™
SDP

Catalog No. NB300516 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300516 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300516 Supplier Novus Biologicals Supplier No. NB300516
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNMT3B Polyclonal antibody specifically detects DNMT3B in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Knockdown.

Specifications

Antigen DNMT3B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ChIP Assay, KnockDown
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2.0 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockdown Validated
Formulation PBS with 1 mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. O88509
Gene Alias DNA (cytosine-5-)-methyltransferase 3 beta, DNA (cytosine-5)-methyltransferase 3B, DNA methyltransferase HsaIIIB, DNA MTase HsaIIIB, Dnmt3b, EC 2.1.1.37, ICF, M.HsaIIIB
Gene Symbols DNMT3B
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the residues M(1) K G D S R H L N E E E G A(14) C of mouse DNMT3b
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 1789
Test Specificity This detects DNA methyltransferase 3b (Dnmt3b) from human and mouse tissues and cells as well as recombinant human Dnmt3b. This detects, to a lesser extent, full-length human recombinant Dnmt3a.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.