Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT3B Rabbit anti-Human, Clone: SY09-07, Novus Biologicals™
SDP

Catalog No. NBP267398 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267398 Supplier Novus Biologicals Supplier No. NBP267398
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

DNMT3B Monoclonal specifically detects DNMT3B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DNMT3B
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone SY09-07
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias DNA (cytosine-5-)-methyltransferase 3 beta, DNA (cytosine-5)-methyltransferase 3B, DNA methyltransferase HsaIIIB, DNA MTase HsaIIIB, Dnmt3b, EC 2.1.1.37, ICF, M.HsaIIIB
Gene Symbols DNMT3B
Host Species Rabbit
Immunogen Synthetic peptide within Human DNMT3B aa 61-110/ 853. (SwissProt: Q9UBC3 Human; SwissProt: O88509 Mouse; Unigene: 117353 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cancer, Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, DNA Repair, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 1789
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.