Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dnmt3L Antibody (5H4A5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
| Antigen | Dnmt3L |
|---|---|
| Clone | 5H4A5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61755
![]() |
Novus Biologicals
NBP261755 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
Dnmt3L Monoclonal specifically detects Dnmt3L in Human samples. It is validated for Western Blot, ELISA.Specifications
| Dnmt3L | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Chromatin Research | |
| 29947 | |
| Purified recombinant fragment of human DNMT3L (AA: 147-386) expressed in E. Coli. | |
| Primary | |
| 43.6 kDa |
| 5H4A5 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Unconjugated | |
| Mouse | |
| DNA (cytosine-5-)-methyltransferase 3-like, DNA (cytosine-5)-methyltransferase 3-like, human cytosine-5-methyltransferase 3-like protein, 10cytosine-5-methyltransferase 3-like protein, MGC1090 | |
| DNMT3L | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title