Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dnmt3L Antibody (5H4A5) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261755 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61755 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61755 Supplier Novus Biologicals Supplier No. NBP261755
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dnmt3L Monoclonal specifically detects Dnmt3L in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Dnmt3L
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 5H4A5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000
Formulation PBS with 0.05% Sodium Azide
Gene Alias DNA (cytosine-5-)-methyltransferase 3-like, DNA (cytosine-5)-methyltransferase 3-like, human cytosine-5-methyltransferase 3-like protein, 10cytosine-5-methyltransferase 3-like protein, MGC1090
Gene Symbols DNMT3L
Host Species Mouse
Immunogen Purified recombinant fragment of human DNMT3L (AA: 147-386) expressed in E. Coli.
Molecular Weight of Antigen 43.6 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 29947
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.