Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dnmt3L Antibody, Novus Biologicals™
SDP

Catalog No. NBP227098 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227098 0.1 mg
NBP227098SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227098 Supplier Novus Biologicals Supplier No. NBP227098
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Dnmt3L Polyclonal specifically detects Dnmt3L in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Dnmt3L
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4ug/ml∼, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24529261), Immunohistochemistry-Paraffin 5ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9UJW3
Gene Alias DNA (cytosine-5-)-methyltransferase 3-like, DNA (cytosine-5)-methyltransferase 3-like, human cytosine-5-methyltransferase 3-like protein, 10cytosine-5-methyltransferase 3-like protein, MGC1090
Gene Symbols DNMT3L
Host Species Rabbit
Immunogen A portion of amino acids 130-180 from human Dnmt3L was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 29947
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, and chimpanzee, 94% in macaque, rhesus monkey, and white-cheeked gibbon, 89% in panda, cow, new world monkey, Chinese hamster, horse, rat, and pig, 83% in dog, and mouse, 78% in naked mole rat, 73% in elephant, and 67% homologous in opossum.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.