Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dnmt3L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$206.50 - $426.00
Specifications
Antigen | Dnmt3L |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 2-4ug/ml∼, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24529261), Immunohistochemistry-Paraffin 5ug/ml |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227098SS
![]() |
Novus Biologicals
NBP227098SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP227098
![]() |
Novus Biologicals
NBP227098 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
Dnmt3L Polyclonal specifically detects Dnmt3L in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Dnmt3L | |
Western Blot 2-4ug/ml∼, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24529261), Immunohistochemistry-Paraffin 5ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Primate | |
Q9UJW3 | |
29947 | |
A portion of amino acids 130-180 from human Dnmt3L was used as the immunogen for this antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Chromatin Research | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
DNA (cytosine-5-)-methyltransferase 3-like, DNA (cytosine-5)-methyltransferase 3-like, human cytosine-5-methyltransferase 3-like protein, 10cytosine-5-methyltransferase 3-like protein, MGC1090 | |
DNMT3L | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen for this antibody is 100% homologous in human, and chimpanzee, 94% in macaque, rhesus monkey, and white-cheeked gibbon, 89% in panda, cow, new world monkey, Chinese hamster, horse, rat, and pig, 83% in dog, and mouse, 78% in naked mole rat, 73% in elephant, and 67% homologous in opossum. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title