Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DNQX disodium salt
GREENER_CHOICESDP

Catalog No. 231210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-121-0 10 mg
23-125-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-121-0 Supplier Tocris Bioscience™ Supplier No. 2312/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective non-NMDA iGluR antagonist; water-soluble salt of DNQX (Cat. No. 0189)

DNQX disodium salt is a more water-soluble form of DNQX (Cat. No. 0189). DNQX is a selective non-NMDA glutamate receptor antagonist. (IC50 = 0.1 μM and 0.5 μM for kainate and AMPA receptors, respectively). DNQX is a neuroleptic agent that shows pro-oxidative properties. DNQX selectively depolarizes rat thalamic reticular nucleus neurons.

Chemical Identifiers

CAS 1312992-24-7
Molecular Formula C8H2N4Na2O6

Specifications

Quantity 10 mg
Formula Weight 296.1
Purity >98%
Molecular Weight (g/mol) 296.106
Chemical Name or Material DNQX disodium salt
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.