Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNTTIP1 Antibody (OTI1F4), FITC, Novus Biologicals™
SDP

Catalog No. NBP272352F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272352F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272352F Supplier Novus Biologicals Supplier No. NBP272352F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNTTIP1 Monoclonal specifically detects DNTTIP1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DNTTIP1
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1F4
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias C20orf167, chromosome 20 open reading frame 167, deoxynucleotidyltransferase terminal-interacting protein 1, deoxynucleotidyltransferase, terminal, interacting protein 1, dJ447F3.4, TDIF1, TdT binding protein, TdT-interacting factor 1, Terminal deoxynucleotidyltransferase-interacting factor 1
Gene Symbols DNTTIP1
Host Species Mouse
Immunogen Full length recombinant protein of human DNTTIP1 (NP_443183) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 116092.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.