Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNTTIP1 Antibody (OTI1F4), PerCP, Novus Biologicals™
SDP

Catalog No. NB288217 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288217 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288217 Supplier Novus Biologicals Supplier No. NBP272352PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNTTIP1 Monoclonal antibody specifically detects DNTTIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen DNTTIP1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone OTI1F4
Conjugate PerCP
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias C20orf167, chromosome 20 open reading frame 167, deoxynucleotidyltransferase terminal-interacting protein 1, deoxynucleotidyltransferase, terminal, interacting protein 1, dJ447F3.4, TDIF1, TdT binding protein, TdT-interacting factor 1, Terminal deoxynucleotidyltransferase-interacting factor 1
Host Species Mouse
Immunogen Full length human recombinant protein of human DNTTIP1 (NP_443183) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 116092
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.