Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOCK10 Antibody, Novus Biologicals™
SDP

Catalog No. NB10060669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10060669 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10060669 Supplier Novus Biologicals Supplier No. NB10060669
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DOCK10 Polyclonal specifically detects DOCK10 in Human, Mouse samples. It is validated for Gene Knock-Out, Immunoprecipitation, Western Blot.

Specifications

Antigen DOCK10
Applications Gene Knock-Out, Immunoprecipitation, Immunoassay
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 ug/mg lysate, Knockout Validated
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q96BY6
Gene Alias dedicator of cytokinesis 10, dedicator of cytokinesis protein 10, DKFZp781A1532, DRIP2, KIAA0694protein zizimin 3, Nbla10300, ZIZ3dopamine receptor interacting protein 2, zizimin3, zizimin-3
Gene Symbols DOCK10
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human dedicator of cytokinesis 10 using the numbering given in entry NP_055504.2 (GeneID 55619).
Molecular Weight of Antigen 249.5 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55619
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.