Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOCK2 Antibody (OTI7G2), Biotin, Novus Biologicals™
SDP

Catalog No. NBP270583B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270583B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270583B Supplier Novus Biologicals Supplier No. NBP270583B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DOCK2 Monoclonal specifically detects DOCK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen DOCK2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7G2
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias dedicator of cytokinesis 2, dedicator of cytokinesis protein 2, FLJ46592, KIAA0209dedicator of cyto-kinesis 2
Gene Symbols DOCK2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1544-1830 of human DOCK2 (NP_004937) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1794.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.