Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOCK7 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB290860 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290860 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290860 Supplier Novus Biologicals Supplier No. NBP178749PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DOCK7 Polyclonal antibody specifically detects DOCK7 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen DOCK7
Applications Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias dedicator of cytokinesis 7, dedicator of cytokinesis protein 7, KIAA1771ZIR2
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 2090 and 2140 of human Dedicator of Cytokinesis 7 using the numbering given in entry Q96N67.4 (GeneID 85440).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, GPCR, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 85440
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.