Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOCK9 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB253935 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253935 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB253935 Supplier Novus Biologicals Supplier No. NB500265DL594

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

DOCK9 Polyclonal antibody specifically detects DOCK9 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen DOCK9
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Cdc42 guanine nucleotide exchange factor zizimin-1, dedicator of cytokinesis 9, dedicator of cytokinesis protein 9, DKFZp686C11110, DKFZp686D2047, DKFZp686N04132, FLJ11949, FLJ16744, FLJ45601, KIAA1058FLJ44528, KIAA1085, ZIZ1FLJ45282, ZIZIMIN1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 1850 and 1900 of human Dedicator of Cytokinesis 9 using the numbering given in entry NP_056111.1 (GeneID 23348).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23348
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.