Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOCK9 Antibody, Novus Biologicals™
SDP

Catalog No. NB500263 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500263 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500263 Supplier Novus Biologicals Supplier No. NB500263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DOCK9 Polyclonal specifically detects DOCK9 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen DOCK9
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:15000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_056111.1
Gene Alias Cdc42 guanine nucleotide exchange factor zizimin-1, dedicator of cytokinesis 9, dedicator of cytokinesis protein 9, DKFZp686C11110, DKFZp686D2047, DKFZp686N04132, FLJ11949, FLJ16744, FLJ45601, KIAA1058FLJ44528, KIAA1085, ZIZ1FLJ45282, ZIZIMIN1
Gene Symbols DOCK9
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Dedicator of Cytokinesis 9 using the numbering given in entry NP_056111.1 (GeneID 23348).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23348
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.