Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOCK9 Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7140253
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-7844 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-7844 Supplier Bethyl Laboratories, Inc Supplier No. A300530AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Formin-like protein 2 is a cytoplasmic phosphoprotein belonging to the formin homology domain containing protein family and has a DAD (diaphanous autoregulatory domain), FH2 (formin homology 2) domain, and GBD/FH3 (Rho GTPase-binding/formin homology 3) domain. The exact function of FMNL2 is yet to be known but reports suggest that it may have a role in the Wnt signaling pathway. Formin-related proteins are also known to play an important role in morphogenesis, cytokinesis and cell polarity. Several alternatively spliced transcript variants have been described but their full-length nature is not yet determined. It is also known to be associated with a diffuse-type gastric cancer, breast cancer, chondrosarcoma, melanoma, and glioblastoma. FMNL2 might be implicated in polarity control, invasion, migration or metastasis through actin filament nucleation and may be regulated by Rho family GTPase.

Specifications

Antigen DOCK9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene DOCK9
Gene Accession No. Q9BZ29
Gene Alias cdc42 guanine nucleotide exchange factor zizimin-1, KIAA1058, ZIZ1, zizimin1
Gene Symbols DOCK9
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23348
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.