Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecylbenzene Sodium Sulfonate, Powder, Approx. 90%, Spectrum™ Chemical
SDP

Catalog No. 18602316 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 g
500 g
3 kg
12 kg
25 kg
90 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

D1064, 25155-30-0, C18H29NaO3S

Spectrum™ Chemical Dodecylbenzene Sodium Sulfonate, Powder

Chemical Identifiers

CAS 25155-30-0
Molecular Formula C18H29NaO3S
Molecular Weight (g/mol) 348.48
InChI Key HFQQZARZPUDIFP-UHFFFAOYSA-M
IUPAC Name sodium 2-dodecylbenzene-1-sulfonate
SMILES [Na+].CCCCCCCCCCCCC1=C(C=CC=C1)S([O-])(=O)=O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 500 g
Percent Purity ∼90%
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.