Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOK1 Rabbit anti-Human, Mouse, Rat, Clone: 6U4U9, Novus Biologicals™
SDP

Catalog No. NB166521 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166521 100 μg
NB166522 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166521 Supplier Novus Biologicals Supplier No. NBP315713100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DOK1 Monoclonal antibody specifically detects DOK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen DOK1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 6U4U9
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias docking protein 1, docking protein 1 (downstream of tyrosine kinase 1), docking protein 1, 62kD (downstream of tyrosine kinase 1), docking protein 1, 62kDa (downstream of tyrosine kinase 1), Downstream of tyrosine kinase 1, MGC117395, MGC138860, p62(dok), P62DOK, pp62
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 382-481 of human DOK1 (Q99704).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1796
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.