Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOK5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $499.50
Specifications
Antigen | DOK5 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP156420UL
![]() |
Novus Biologicals
NBP15641120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP156411
![]() |
Novus Biologicals
NBP156411 |
100 μL |
Each for $499.50
|
|
|||||
Description
DOK5 Polyclonal specifically detects DOK5 in Human samples. It is validated for Western Blot.Specifications
DOK5 | |
Polyclonal | |
Rabbit | |
Q9P104 | |
55816 | |
Synthetic peptides corresponding to DOK5(docking protein 5) The peptide sequence was selected from the N terminal of DOK5 (NP_060901). Peptide sequence GPKRLEKFSDERAAYFRCYHKVTELNNVKNVARLPKSTKKHAIGIYFNDD. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
C20orf180, chromosome 20 open reading frame 180, dJ805C22.1, docking protein 5, Downstream of tyrosine kinase 5, Insulin receptor substrate 6, IRS6, IRS-6, MGC16926 | |
DOK5 | |
IgG | |
34 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title