Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOK7 Antibody (OTI2C12), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP272491A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272491A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272491A30 Supplier Novus Biologicals Supplier No. NBP272491AF350

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

DOK7 Monoclonal specifically detects DOK7 in Human samples. It is validated for Western Blot.

Specifications

Antigen DOK7
Applications Western Blot
Classification Monoclonal
Clone OTI2C12
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C4orf25chromosome 4 open reading frame 25, CMS1B, docking protein 7, Dok-7, Downstream of tyrosine kinase 7, FLJ33718, FLJ39137, FLJ90556, protein Dok-7
Gene Symbols DOK7
Host Species Mouse
Immunogen Full length recombinant protein of human DOK7 (NP_775931) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 285489
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.