Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOK7 Antibody (OTI2C12), PE, Novus Biologicals™
SDP

Catalog No. NB288105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288105 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288105 Supplier Novus Biologicals Supplier No. NBP272491PE

Mouse Monoclonal Antibody

DOK7 Monoclonal antibody specifically detects DOK7 in Human samples. It is validated for Western Blot

Specifications

Antigen DOK7
Applications Western Blot
Classification Monoclonal
Clone OTI2C12
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias C4orf25chromosome 4 open reading frame 25, CMS1B, docking protein 7, Dok-7, Downstream of tyrosine kinase 7, FLJ33718, FLJ39137, FLJ90556, protein Dok-7
Host Species Mouse
Immunogen Full length human recombinant protein of human DOK7(NP_775931) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285489
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.