Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Donepezil hydrochloride
GREENER_CHOICE

Catalog No. p-4817074
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent AChE inhibitor

Potent acetylcholinesterase inhibitor (IC50 = 5.7 nM). Displays 1252-fold selectivity for AChE over BuChE (IC50 = 7138 nM for BuChE). Orally active.

Deuterated analog also available.

Chemical Identifiers

CAS 120011-70-3
Molecular Formula C24H30ClNO3

Specifications

Quantity 10 mg
Formula Weight 415.95
Purity >99%
Molecular Weight (g/mol) 415.96
Chemical Name or Material Donepezil hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.