Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Dooku 1

Click to view available options
Quantity:
10 mg
50 mg
Description
Dooku 1 reversibly blocks Yoda1 (Cat. No. 5586) activity (IC50 = ∼1.5 μM). Exhibits no effect on constitutive Piezo1 channel activity, but antagonizes Yoda1-induced Piezo1 activation in endothelial cells, and inhibits Yoda1-induced Ca2+-entry in vitro. Also blocks Yoda1-induced relaxation of aorta ex vivo.
Specifications
Specifications
Chemical Name or Material | 2-[(2,6-Dichlorobenzyl)thio)-5-(1 H-pyrrol-2-yl)-1,3,4-oxadiazole |
CAS | 2253744-54-4 |
Quantity | 10 mg |
Target | Other Piezo Channel Compounds |
Molecular Formula | C13H9Cl2N3OS |
Formula Weight | 326.2 |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction