Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopa Decarboxylase/DDC Antibody, Novus Biologicals™
SDP

Catalog No. NB300252 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300252 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300252 Supplier Novus Biologicals Supplier No. NB300252
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dopa Decarboxylase/DDC Polyclonal specifically detects Dopa Decarboxylase/DDC in Human, Rat, Bovine, Canine, Guinea Pig, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Dopa Decarboxylase/DDC
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. P20711
Gene Alias AADCaromatic-L-amino-acid decarboxylase, DOPA decarboxylase, dopa decarboxylase (aromatic L-amino acid decarboxylase), EC 4.1.1, EC 4.1.1.28
Gene Symbols DDC
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acid residues from the N-terminal region conjugated to KLH. Accession # P20711
Molecular Weight of Antigen 55 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1644
Test Specificity Specific for the ∼55k DDC protein.
Target Species Human, Rat, Bovine, Canine, Guinea Pig, Rabbit, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.