Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D1R/DRD1 Antibody (SG2-D1a), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB11060017W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11060017W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11060017W Supplier Novus Biologicals Supplier No. NB11060017AF405

Mouse Monoclonal Antibody

Dopamine D1R/DRD1 Monoclonal specifically detects Dopamine D1R/DRD1 in Mouse, Rat, Bovine (Negative), Canine (Negative), Human (Negative), Porcine (Negative), Rabbit (Negative), Sheep (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.

Specifications

Antigen Dopamine D1R/DRD1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating)
Classification Monoclonal
Clone SG2-D1a
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias D(1A) dopamine receptor, D1DR, DADR, Dopamine D1 receptor, dopamine receptor D1, DRD1A
Gene Symbols DRD1
Host Species Mouse
Immunogen Recombinant rat Dopamine Receptor D1 protein near the C-terminus. [Swiss-Prot# P18901]
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 1812
Test Specificity This antibody is specific for Dopamine Receptor D1A. There is no cross-reactivity with D1B (D5) receptor.
Reconstitution Protein A purified
Target Species Mouse, Rat, Bovine (Negative), Canine (Negative), Human (Negative), Porcine (Negative), Rabbit (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.